missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzonitrile hydrochloride, 97%

Product Code. 11387349
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387349 25 g 25g
11377349 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387349 Supplier Thermo Scientific Alfa Aesar Supplier No. H27473.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15996-76-6 | C8H9ClN2 | 168.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15996-76-6
Molecular Formula C8H9ClN2
Molecular Weight (g/mol) 168.624
MDL Number MFCD01861472
InChI Key QREZLLYPLRPULF-UHFFFAOYSA-N
Synonym 4-aminomethyl benzonitrile hydrochloride, 4-cyanobenzylamine hcl, 4-cyanobenzylamine hydrochloride, 4-aminomethyl benzonitrile hcl, 4-aminomethyl-benzonitrile hydrochloride, benzonitrile, 4-aminomethyl-, monohydrochloride, p-aminomethyl benzonitrile hydrochloride, pubchem2057, acmc-209dkg, 4-aminomethylbenzonitrile hcl
PubChem CID 9942250
IUPAC Name 4-(aminomethyl)benzonitrile;hydrochloride
SMILES C1=CC(=CC=C1CN)C#N.Cl

Specifications

Melting Point 279°C to 284°C
Quantity 25 g
Formula Weight 168.62
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)benzonitrile hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.