missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethylphenylboronic acid hydrochloride, 96%

Product Code. 10629102 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10629102 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10629102 Supplier Thermo Scientific Acros Supplier No. 358842500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75705-21-4 | C7H11BClNO2 | 187.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 75705-21-4
CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C7H11BClNO2
MDL Number MFCD01632199
Quantity 250 mg
Synonym 4-aminomethylphenylboronic acid hydrochloride, 4-aminomethyl phenylboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hcl, 4-aminomethyl benzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid hcl, 4-aminomethylphenyl boronic acid hydrochloride, 4-aminomethyl phenyl boronic acid hydrochloride, 4-aminomethylbenzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hydrochloride
InChI Key HUZNRXFJHYNUMV-UHFFFAOYSA-N
SMILES Cl.NCC1=CC=C(C=C1)B(O)O
IUPAC Name [4-(aminomethyl)phenyl]boronic acid;hydrochloride
Molecular Weight (g/mol) 187.43
PubChem CID 2734311
Formula Weight 187.43
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Aminomethylphenylboronic acid hydrochloride, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminomethylphenylboronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.