missing translation for 'onlineSavingsMsg'
Learn More

(4-Aminophenylthio)acetic acid, 97%, Thermo Scientific™

Product Code. 11498677
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11498677

Brand: Thermo Scientific Alfa Aesar A19834.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It can react with 2-phenyl-benzo[d][1,3]oxazin-4-one to get 2-(Phenyl-4-(3H)-quinazolyl)-phenyl-4′-mercaptoacetic acid. This reaction needs solvents pyridine and H2O by heating.

Solubility
Soluble in methanol, acetone, toluene, n-octanol, acetonitrile, ethyl acetate, dichloromethane.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-18-7
Molecular Formula C8H9NO2S
Molecular Weight (g/mol) 183.225
MDL Number MFCD00025360
InChI Key CTPIHHXCACYCIV-UHFFFAOYSA-N
Synonym 4-aminophenylthio acetic acid, 2-4-aminophenylthio acetic acid, 4-aminophenyl sulfanyl acetic acid, acetic acid, 4-aminophenyl thio, p-aminophenylthioacetic acid, p-aminophenylmercaptoacetic acid, acetic acid,2-4-aminophenyl thio, p-aminothiophenoxyacetic acid, acetic acid, p-aminophenyl thio
PubChem CID 66907
IUPAC Name 2-(4-aminophenyl)sulfanylacetic acid
SMILES C1=CC(=CC=C1N)SCC(=O)O

Specifications

Melting Point ∼200°C (decomposition)
Quantity 1 g
Beilstein 2720454
Solubility Information Soluble in methanol,acetone,toluene,n-octanol,acetonitrile,ethyl acetate,dichloromethane.
Formula Weight 183.23
Percent Purity 97%
Chemical Name or Material (4-Aminophenylthio)acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.