missing translation for 'onlineSavingsMsg'
Learn More

4-Aminosalicylic acid, 98+%

Product Code. 11440897
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440897 500 g 500g
11430897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440897 Supplier Thermo Scientific Alfa Aesar Supplier No. B23289.36
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65-49-6 | C7H7NO3 | 153.137 g/mol

Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.

Solubility
Soluble in water (2 g/L) at 20°C, dilute sodium hydroxide, dilute nitric acid, ether (slightly), methanol, acetone, and ethanol. Virtually insoluble in benzene, chloroform, and carbon tetrachloride.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-49-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007789
InChI Key WUBBRNOQWQTFEX-UHFFFAOYSA-N
Synonym 4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil
PubChem CID 4649
ChEBI CHEBI:27565
IUPAC Name 4-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)O)C(=O)O

Specifications

Melting Point ∼135°C to 145°C (decomposition)
Odor Odorless
Quantity 500 g
Beilstein 473071
Merck Index 14,477
Solubility Information Soluble in water (2g/L) at 20°C,dilute sodium hydroxide,dilute nitric acid,ether (slightly),methanol,acetone,and ethanol. Virtually insoluble in benzene,chloroform,and carbon tetrachloride.
Formula Weight 153.14
Percent Purity ≥98%
Chemical Name or Material 4-Aminosalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.