missing translation for 'onlineSavingsMsg'
Learn More

4-Aminosalicylic Acid, TRC

Product Code. 30012686
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012686 5 g 5g
30009954 10 g 10g
17980685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012686 Supplier Toronto Research Chemicals Supplier No. TRCA6292205G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-49-6
Molecular Formula C7 H7 N O3
Molecular Weight (g/mol) 153.14
Synonym Salicylic acid, 4-amino- (8CI), 4-Amino-2-hydroxybenzoic acid, 2-Hydroxy-4-aminobenzoic acid, 3-Hydroxy-4-carboxyaniline, 4-ASA, 4-Aminosalicylic acid, 4-Carboxy-3-hydroxyaniline, Amino-PAS, Apacil, Apas, Deapasil, Entepas, Euteropas, Gabbropas, Monopass, NSC 2083, NSC 211698, Osacyl, PAS, PAS (acid), PASK, Pamacyl, Pamisyl, Para-Pas, Para-aminosalicylic acid, Paramycin, Parasal, Parasalicil, Parasalindon, Pasa, Pasalon, Pasara, Pasem, Paser, Pasmed, Pasnodia, Pasolac, Propasa, Rezipas, Sanipirol-4, p-Aminosalicylic acid
IUPAC Name 4-amino-2-hydroxybenzoic acid
SMILES Nc1ccc(C(=O)O)c(O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H7NO3/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,9H,8H2,(H,10,11)
Formula Weight 153.0426
Percent Purity >95%
Chemical Name or Material 4-Aminosalicylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.