missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoveratrole, 98%

Product Code. 10656092 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656092 25 g Glass bottle 25g
10122800 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656092 Supplier Thermo Scientific Acros Supplier No. 104720250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6315-89-5 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6315-89-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008394
InChI Key LGDHZCLREKIGKJ-UHFFFAOYSA-N
Synonym 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
PubChem CID 22770
IUPAC Name 3,4-dimethoxyaniline
SMILES COC1=C(C=C(C=C1)N)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 88°C
Color Brown
Boiling Point 174°C to 176°C (22.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H3(OCH3)2
Quantity 25 g
Beilstein 13, 780
Solubility Information Solubility in water: insoluble
Formula Weight 153.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoveratrole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Aminoveratrole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.