missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoyl-4'-bromobiphenyl, 99%

Product Code. 11428397
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428397 25 g 25g
11438397 100 g 100g
11448397 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428397 Supplier Thermo Scientific Alfa Aesar Supplier No. A18482.14
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 63242-14-8 | C19H13BrO | 337.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63242-14-8
Molecular Formula C19H13BrO
Molecular Weight (g/mol) 337.216
MDL Number MFCD00209630
InChI Key ITMLYLPKSFZCJK-UHFFFAOYSA-N
Synonym 4-benzoyl-4'-bromobiphenyl, 4-4-bromophenyl benzophenone, 4'-bromobiphenyl-4-yl phenyl methanone, 4'-bromo-1,1'-biphenyl-4-yl phenyl methanone, 4-4-bromophenyl phenyl-phenyl-methanone, 4-4-bromophenyl phenyl phenyl ketone, 4-4-bromophenyl phenyl-phenylmethanone, 4-4-bromophenyl phenyl phenyl methanone, pubchem9089
PubChem CID 478901
IUPAC Name [4-(4-bromophenyl)phenyl]-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 155°C to 158°C
Quantity 25 g
Beilstein 2378429
Formula Weight 337.22
Percent Purity 99%
Chemical Name or Material 4-Benzoyl-4'-bromobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.