missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbenzoic acid, 98%

Product Code. 11497207
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497207 5 g 5g
11487207 1 g 1g
11407217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497207 Supplier Thermo Scientific Alfa Aesar Supplier No. A14993.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-95-0 | C14H10O3 | 226.231 g/mol

4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
It is soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Incompatible with strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-95-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00002560
InChI Key IFQUPKAISSPFTE-UHFFFAOYSA-N
Synonym p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
PubChem CID 69147
IUPAC Name 4-benzoylbenzoic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 198°C to 200°C
Odor Odorless
Quantity 5 g
Beilstein 1960224
Formula Weight 226.23
Percent Purity 99%
Chemical Name or Material 4-Benzoylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.