missing translation for 'onlineSavingsMsg'
Learn More

(4-Benzyl-1,4-oxazinan-2-yl)methanol, Thermo Scientific™

Product Code. 10561692
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10561692 1 g 1g
10101262 250 mg 250mg
10280395 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10561692 Supplier Thermo Scientific Maybridge Supplier No. CC12409DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40987-24-4
Molecular Formula C12H18NO2
Molecular Weight (g/mol) 208.28
MDL Number MFCD02682009,MFCD10698663,MFCD11848685
InChI Key WQNIKIMRIXHNFF-GFCCVEGCSA-O
Synonym 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw
PubChem CID 2776358
SMILES OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (4-benzylmorpholin-2-yl)methanol
Formula Weight 207.27
Percent Purity 95%
Chemical Name or Material (4-Benzyl-1, 4-oxazinan-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.