missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)-2-hydroxy-5-(sulfooxy)benzoic Acid, TRC

Product Code. 30329507
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329507 100 mg 100mg
30346747 250 mg 250mg
30343476 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30329507 Supplier Toronto Research Chemicals Supplier No. TRCB279285100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyloxy)-2-hydroxy-5-(sulfooxy)benzoic Acid
Analyte or Component Names 4-(Benzyloxy)-2-hydroxy-5-(sulfooxy)benzoic Acid
Shipping Condition Room Temperature
Molecular Formula C14 H12 O8 S
InChI Formula InChI=1S/C14H12O8S/c15-11-7-12(21-8-9-4-2-1-3-5-9)13(22-23(18,19)20)6-10(11)14(16)17/h1-7,15H,8H2,(H,16,17)(H,18,19,20)
Solution Type Neat
SMILES OC(=O)c1cc(OS(=O)(=O)O)c(OCc2ccccc2)cc1O
IUPAC Name 2-hydroxy-4-phenylmethoxy-5-sulfooxybenzoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 340.305
Quantity 100 mg
Formula Weight 340.025
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.