missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)phenol, 98%

Product Code. 10791631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10791631 50 g Glass bottle 50g
10457010 250 g Glass bottle 250g
13498408 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10791631 Supplier Thermo Scientific Acros Supplier No. 169780500
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 103-16-2 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121.0°C to 122.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4OH
Quantity 50 g
Beilstein 06,845
Merck Index 15,6334
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,benzene,diethyl ether,,soluble in acetone and alkalies,,practically insoluble in petroleum hydrocarbons
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 4-(Benzyloxy)phenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxyphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.