missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybromobenzene, 97%

Product Code. 11944711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11944711 50 g 50g
11934711 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11944711 Supplier Thermo Scientific Acros Supplier No. 442460500
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6793-92-6 | C13H11BrO | 263.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6793-92-6
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.14
InChI Key OUQSGILAXUXMGI-UHFFFAOYSA-N
Synonym 4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene
PubChem CID 138835
IUPAC Name 1-bromo-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)Br

Specifications

Melting Point 62°C to 64°C
Boiling Point 166°C to 167°C (4.0 Torr)
Assay Percent Range 96% min. (GC)
Quantity 50 g
Formula Weight 263.14
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybromobenzene
compliance-icons
Product Identifier
  • 4-Benzyloxybromobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.