missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenol, 98+%

Product Code. 11432617
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
1000 g
Unit Size:
1000g
100g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432617 1000 g 1000g
11329429 25 g 25g
15463097 50 g 50g
11339429 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11432617 Supplier Thermo Scientific Alfa Aesar Supplier No. A14843.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-16-2 | C13H12O2 | 200.237 g/mol

4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol

Specifications

Melting Point 118°C to 122°C
Density 1.26
Quantity 1000 g
Beilstein 1958305
Merck Index 14,6248
Solubility Information Slightly soluble in water. Soluble in alcohol,acetone,benzene,and diethyl ether.
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O
Molecular Weight (g/mol) 200.237
Formula Weight 200.24
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.