missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenylacetonitrile, 98+%, Thermo Scientific™

Product Code. 10473575 Shop All Thermo Scientific Products
Click to view available options
:
5g; Glass bottle
This item is not returnable. View return policy

Product Code. 10473575

Brand: Thermo Scientific 394060050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 838-96-0
CAS Min % 98.0
CAS Max % 100.0
Melting Point 65°C to 67°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C15H13NO
Linear Formula C6H5CH2OC6H4CH2CN
MDL Number MFCD00016400
Quantity 5 g
Synonym 4-benzyloxyphenylacetonitrile, 2-4-benzyloxy phenyl acetonitrile, 4-benzyloxybenzyl cyanide, 4-benzyloxy phenyl acetonitrile, 2-4-phenylmethoxy phenyl ethanenitrile, acmc-20amoi, pubchem15926, 4-benzyloxybenzeneacetonitrile, p-benzyloxyphenyl acetonitrile
InChI Key QKEYZRVDFZDOEP-UHFFFAOYSA-N
SMILES N#CCC1=CC=C(OCC2=CC=CC=C2)C=C1
IUPAC Name 2-(4-phenylmethoxyphenyl)acetonitrile
Molecular Weight (g/mol) 223.28
PubChem CID 522804
Formula Weight 223.27
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Benzyloxyphenylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxyphenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.