missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Benzylphenol, 98+%

Product Code. 11405107
Click to view available options
Quantity:
25g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11405107

Brand: Alfa Aesar™ B22538.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 101-53-1
Boiling Point 320°C to 322°C
Molecular Formula C13H12O
MDL Number MFCD00002384
Quantity 25g
Beilstein 1449572
Merck Index 14,1143
Synonym p-benzylphenol, fesiasept, 4-hydroxydiphenylmethane, 4-hydroxyditane, 4-phenylmethyl phenol, phenol, 4-phenylmethyl, p-hydroxydiphenyl methane, alpha-phenyl-p-cresol, p-hydroxydiphenylmethane, 4-benzyl-phenol
InChI Key HJSPWKGEPDZNLK-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)O
IUPAC Name 4-benzylphenol
Molecular Weight (g/mol) 184.238
PubChem CID 7563
Formula Weight 184.24
Percent Purity ≥98%
Chemical Name or Material 4-Benzylphenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.