missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylthiomorpholine 1,1-dioxide, 97%, Thermo Scientific™

Product Code. 10314242
Change view
Click to view available options
:
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10314242 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10314242 Supplier Thermo Scientific Maybridge Supplier No. DFP00030EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 26475-66-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H15NO2S
MDL Number MFCD00084894
Quantity 10 g
Synonym 4-benzylthiomorpholine 1,1-dioxide, 1,1-dioxide-4-phenylmethyl thiomorpholine, 4-benzyl-1??-thiomorpholine-1,1-dione, 4-benzylthiomorpholine1,1-dioxide, acmc-209grq, maybridge1_003205, 4-benzylthiomorpholine-1,1-dioxide, 4-benzylthiomorpholine-1,1,-dioxide, 4-benzyl-1,4-thiazaperhydroine-1,1-dione
InChI Key KFAMTQFKYUXQKV-UHFFFAOYSA-N
SMILES O=S1(=O)CCN(CC2=CC=CC=C2)CC1
IUPAC Name 4-benzyl-1,4-thiazinane 1,1-dioxide
Molecular Weight (g/mol) 225.31
PubChem CID 318454
Formula Weight 225.31
Percent Purity 97%
Chemical Name or Material 4-Benzylthiomorpholine 1, 1-dioxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.