missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)phenol, 97%

Product Code. 15481958
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15481958 5 g 5g
15491958 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 15481958

Brand: Thermo Scientific Alfa Aesar H55645.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54840-15-2 | C11H15NO3 | 209.245 g/mol

4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54840-15-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD00226573
InChI Key YRQMBQUMJFVZLF-UHFFFAOYSA-N
Synonym 4-n-boc-aminophenol, tert-butyl 4-hydroxyphenyl carbamate, tert-butyl n-4-hydroxyphenyl carbamate, tert-butyl 4-hydroxyphenylcarbamate, 4-boc-amino phenol, n-boc-4-hydroxyaniline, 1,1-dimethylethyl 4-hydroxyphenyl carbamate, 4-tert-butoxycarbonylamino phenol, 4-hydroxy-phenyl-carbamic acid tert-butyl ester, carbamic acid, 4-hydroxyphenyl-, 1,1-dimethylethyl ester
PubChem CID 2756771
IUPAC Name tert-butyl N-(4-hydroxyphenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)O

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.