missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)phenol, 97%

Product Code. 15491958
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491958 25 g 25g
15481958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491958 Supplier Thermo Scientific Alfa Aesar Supplier No. H55645.14
 Restricted Item
Estimated Shipment: 11-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 54840-15-2 | C11H15NO3 | 209.245 g/mol

4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 54840-15-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD00226573
InChI Key YRQMBQUMJFVZLF-UHFFFAOYSA-N
Synonym 4-n-boc-aminophenol, tert-butyl 4-hydroxyphenyl carbamate, tert-butyl n-4-hydroxyphenyl carbamate, tert-butyl 4-hydroxyphenylcarbamate, 4-boc-amino phenol, n-boc-4-hydroxyaniline, 1,1-dimethylethyl 4-hydroxyphenyl carbamate, 4-tert-butoxycarbonylamino phenol, 4-hydroxy-phenyl-carbamic acid tert-butyl ester, carbamic acid, 4-hydroxyphenyl-, 1,1-dimethylethyl ester
PubChem CID 2756771
IUPAC Name tert-butyl N-(4-hydroxyphenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)O

Spezifikation

Melting Point 143°C to 147°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)phenol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.