missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15498026
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498026 1 g 1g
15408036 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498026 Supplier Thermo Scientific Alfa Aesar Supplier No. H62948.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1351501-44-4 | Molecular Formula: C18H27BFNO4 | Molecular Weight: 351.225 g/mol

4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid generally used in the synthesis of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid generally used in the synthesis of organic compounds.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1351501-44-4
Molecular Formula C18H27BFNO4
Molecular Weight (g/mol) 351.225
InChI Key SAUVAJYHMCIQNE-UHFFFAOYSA-N
Synonym 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester
PubChem CID 66980292
IUPAC Name tert-butyl N-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CNC(=O)OC(C)(C)C)F

Spezifikation

Quantity 1 g
Formula Weight 351.22
Percent Purity 96%
Chemical Name or Material 4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.