missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-naphthoic acid, 98%

Product Code. 15442199
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15442199 5 g 5g
15432199 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15442199

Brand: Thermo Scientific Alfa Aesar H64820.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16650-55-8 | C11H7BrO2 | 251.079 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16650-55-8
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD00094221
InChI Key FIJIPZQZVLCOMB-UHFFFAOYSA-N
Synonym 4-bromo-1-naphthoic acid, 4-bromo-1-naphthalenecarboxylic acid, 4-bromo-1-naphtalenecarboxylic acid, 1-naphthalenecarboxylic acid, 4-bromo, 4-bromo naphthoic acid, acmc-209dum, 4-bromo-naphthalic acid, 4-bromo-1-naphthoicacid, ksc175i3p, 1-naphthalenecarboxylicacid,4-bromo
PubChem CID 282591
IUPAC Name 4-bromonaphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)C(=O)O

Specifications

Quantity 5 g
Beilstein 1950351
Solubility Information Slightly soluble in water.
Formula Weight 251.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-1-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.