missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,2-diphenylbutyronitrile, 95%

Product Code. 11493378
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493378 100 g 100g
11483378 25 g 25g
11403388 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493378 Supplier Thermo Scientific Alfa Aesar Supplier No. A17786.22
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39186-58-8 | C16H14BrN | 300.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39186-58-8
Molecular Formula C16H14BrN
Molecular Weight (g/mol) 300.199
MDL Number MFCD00001845
InChI Key IGYSFJHVFHNOEI-UHFFFAOYSA-N
Synonym 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile
PubChem CID 96575
IUPAC Name 4-bromo-2,2-diphenylbutanenitrile
SMILES C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2

Specifications

Melting Point 67°C to 69°C
Quantity 100 g
UN Number UN3439
Beilstein 1684952
Formula Weight 300.2
Percent Purity 95%
Chemical Name or Material 4-Bromo-2,2-diphenylbutyronitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.