missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3,5,6-tetrafluorobenzoic acid, Thermo Scientific™

Product Code. 10535850
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10535850 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10535850 Supplier Thermo Scientific Maybridge Supplier No. MAY00175DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 4707-24-8
Packaging Amber Glass Bottle
Molecular Formula C7HBrF4O2
Quantity 5 g
Synonym 4-bromo-2,3,5,6-tetrafluoro-benzoic acid, acmc-1arsf, 4-bromotetrafluorobenzoic acid, 4-bromotetrafluorobenzene carboxylic acid, benzoicacid, 4-bromo-2,3,5,6-tetrafluoro, benzoic acid, 4-bromo-2,3,5,6-tetrafluoro, 4-bromo-2,3,5,6-tetrafluorobenzoic acid, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoic acid
InChI Key MTZLICDXLGQWJV-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)O
IUPAC Name 4-bromo-2,3,5,6-tetrafluorobenzoic acid
Molecular Weight (g/mol) 272.981
PubChem CID 298173
Formula Weight 272.98
Percent Purity 95%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.