missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-dichlorophenol, Dr. Ehrenstorfer

Product Code. 30326945
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326945 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326945 Supplier Dr. Ehrenstorfer Supplier No. DREC10726800
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromo-2,5-dichlorophenol
CAS 1940-42-7
Analyte or Component Names 4-Bromo-2,5-dichlorophenol
Shipping Condition +20°C
Molecular Formula C6H3BrCl2O
Certifications/Compliance ISO/IEC 17025
Synonym 2,5-Dichloro-4-bromophenol, Leptophos Phenol, Phosvel Phenol
SMILES Oc1cc(Cl)c(Br)cc1Cl
Molecular Weight (g/mol) 241.9
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.