missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-6-methylphenol, 97%, Thermo Scientific™

Product Code. 10039561
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10039561 1 g 1g
10223662 10 g 10g
10435712 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039561 Supplier Thermo Scientific Maybridge Supplier No. SB01216DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7530-27-0
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00002167
InChI Key IDDUDPYBPXKGCP-UHFFFAOYSA-N
PubChem CID 82031
IUPAC Name 4-bromo-2-chloro-6-methylphenol
SMILES CC1=CC(Br)=CC(Cl)=C1O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3C6H2(OH)(Cl)Br
Quantity 1 g
Synonym 4-bromo-6-chloro-o-cresol, o-cresol, 4-bromo-6-chloro, 4-bromo-6-chloro-2-methylphenol, phenol, 4-bromo-2-chloro-6-methyl, 2-chloro-4-bromo-6-methylphenol, 4-bromo-2-chloro-6-methyl-phenol, 5-bromo-3-chloro-2-hydroxytoluene, #, phenol,4-bromo-2-chloro-6-methyl
Formula Weight 221.48
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-chloro-6-methylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.