missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloroaniline, 99%

Product Code. 10448740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10448740 10 g 10g
10721441 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10448740 Supplier Thermo Scientific Acros Supplier No. 167480100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38762-41-3 | C6H5BrClN | 206.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38762-41-3
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.47
MDL Number MFCD00007660
InChI Key INMZDDDQLHKGPF-UHFFFAOYSA-N
Synonym 2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
PubChem CID 610169
IUPAC Name 4-bromo-2-chloroaniline
SMILES C1=CC(=C(C=C1Br)Cl)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 67°C to 70°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H3(Cl)NH2
Quantity 10 g
Beilstein 12, 652
Formula Weight 206.47
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-chloroaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-chloroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.