missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorophenol, 99%

Product Code. 10433975 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10433975 5 g Glass bottle 5g
10448320 25 g Glass bottle 25g
10523161 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10433975 Supplier Thermo Scientific Acros Supplier No. 173530050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3964-56-5 | C6H4BrClO | 207.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3964-56-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD00002166
InChI Key VIBJPUXLAKVICD-UHFFFAOYSA-N
Synonym 2-chloro-4-bromophenol, phenol, 4-bromo-2-chloro, phenol, 4-bromochloro, unii-994sj34d8p, 4-bromo-2-chloro-phenol, attercop-chm at106179, 4-bromo-2-chlorophenol 25gr, 4-bromo-chlorophenol, pubchem3596, acmc-209j6p
PubChem CID 19859
ChEBI CHEBI:38852
IUPAC Name 4-bromo-2-chlorophenol
SMILES OC1=CC=C(Br)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47°C to 51°C
Color White
Boiling Point 232°C to 235°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)OH
Quantity 5 g
Beilstein 06, II, 187
Formula Weight 207.45
Percent Purity 99%
Physical Form Low Melting Mass or Crystalline Needles, Powder and Chunks
Chemical Name or Material 4-Bromo-2-chlorophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-chlorophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.