missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-ethyl-1-iodobenzene, 97%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11430531
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430531 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430531 Supplier Thermo Scientific Alfa Aesar Supplier No. B20052.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 175278-30-5
Molecular Formula C8H8BrI
Molecular Weight (g/mol) 310.96
MDL Number MFCD00221458
InChI Key JWKQXPHYKRQLEJ-UHFFFAOYSA-N
PubChem CID 2761396
IUPAC Name 4-bromo-2-ethyl-1-iodobenzene
SMILES CCC1=C(C=CC(=C1)Br)I

Specifications

Boiling Point 84°C to 86°C (0.4 mmHg)
Quantity 10 g
Sensitivity Light sensitive
Synonym 4-bromo-2-ethyliodobenzene, benzene,4-bromo-2-ethyl-1-iodo, benzene, 4-bromo-2-ethyl-1-iodo, 4-bromo-2-ethyl-1-iodo-benzene, pubchem3773, acmc-1c9j2, 4-bromo-2-ethyl-iodobenzene, 1-iodo-2-ethyl-4-bromobenzene, 4-bromo-2-ethyl-1-iodobenzene, 4-bromanyl-2-ethyl-1-iodanyl-benzene
Formula Weight 310.97
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-ethyl-1-iodobenzene, stabilized with copper

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.