missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoro-6-nitroanisole, 97%

Product Code. p-7030544
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15483887

Brand: Thermo Scientific Alfa Aesar H30220.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74266-66-3 | C7H5BrFNO3 | 250.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74266-66-3
Molecular Formula C7H5BrFNO3
Molecular Weight (g/mol) 250.02
MDL Number MFCD00013374
InChI Key CVCAYLYLJKBSNV-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoro-6-nitroanisole, 5-bromo-3-fluoro-2-methoxy-1-nitrobenzene, 5-bromo-1-fluoro-2-methoxy-3-nitro-benzene, benzene, 5-bromo-1-fluoro-2-methoxy-3-nitro, 4-bromo-2-fluoro-6-nitrophenyl methyl ether, timtec-bb sbb005948, acmc-20amnn, pubchem4134, 4-bromo-2-fluoro-6-nitroanisol
PubChem CID 557159
IUPAC Name 5-bromo-1-fluoro-2-methoxy-3-nitrobenzene
SMILES COC1=C(F)C=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 53°C to 56°C
Quantity 5 g
Formula Weight 250.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluoro-6-nitroanisole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.