missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzaldehyde, 97%

Product Code. 11453147
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11453147

Brand: Thermo Scientific Alfa Aesar A16112.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57848-46-1 | C7H4BrFO | 203.01 g/mol

4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.

Solubility
Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57848-46-1
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00143261
InChI Key UPCARQPLANFGQJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde
PubChem CID 143517
IUPAC Name 4-bromo-2-fluorobenzaldehyde
SMILES FC1=CC(Br)=CC=C1C=O

Specifications

Melting Point 59°C to 63°C
Boiling Point 42°C (19 mmHg)
Quantity 5 g
Beilstein 7700208
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 203.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluorobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.