missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzyl bromide, 99%, Thermo Scientific™

Product Code. 10798532 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10798532

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 76283-09-5
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD00055467
InChI Key XMHNLZXYPAULDF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl bromide, 4-bromo-1-bromomethyl-2-fluorobenzene, 2-fluoro-4-bromobenzyl bromide, 4-bromo-2-fluorobenzylbromide, 2-fluoro-4-bromo-benzylbromide, alpha,4-dibromo-2-fluorotoluene, unii-0z26xyc1jy, 4,alpha-dibromo-2-fluorotoluene, 4-bromo-1-bromomethyl-2-fluoro-benzene, 0z26xyc1jy
PubChem CID 2733660
IUPAC Name 4-bromo-1-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >30.0°C
Color White
Boiling Point 126.0°C (19.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 5 g
Formula Weight 267.92
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 4-Bromo-2-fluorobenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.