missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorophenylacetic acid, 98%

Product Code. 15477678
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477678 25 g 25g
15467678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477678 Supplier Thermo Scientific Alfa Aesar Supplier No. H61176.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 114897-92-6 | C8H6BrFO2 | 233.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114897-92-6
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD09032952
InChI Key PNBIYFPZODYMOO-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorophenylacetic acid, 2-4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluoro-phenyl-acetic acid, 2-4-bromo-2-fluoro-phenyl acetic acid, 4-bromo-2-fluorophenylaceticacid, pubchem13742, acmc-1bskq, ksc493s2b, 4-bromo-2-fluoro-phenylacetic acid
PubChem CID 14053792
IUPAC Name 2-(4-bromo-2-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1Br)F)CC(=O)O

Specifications

Quantity 25 g
Formula Weight 233.03
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-fluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.