missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-iodoaniline, 97%

Product Code. 10451304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10451304 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10451304 Supplier Thermo Scientific Acros Supplier No. 397910050
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 66416-72-6 | C6H5BrIN | 297.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 66416-72-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 69°C to 72°C
Color Gray to Purple
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H5BrIN
Linear Formula BrC6H3(I)NH2
MDL Number MFCD02093764
Quantity 5 g
Synonym 4-bromo-2-iodo-phenylamine, 4-bromo-2-iodobenzenamine, 2-iodo-4-bromoaniline, benzenamine,4-bromo-2-iodo, benzenamine, 4-bromo-2-iodo, pubchem3784, acmc-1b9hb, 4-bromo-2-iodophenylamine, asischem t54343, 4-bromo-2-iodo-benzenamine
InChI Key HHTYEQWCHQEJNV-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Br)I)N
IUPAC Name 4-bromo-2-iodoaniline
Molecular Weight (g/mol) 297.91
PubChem CID 823934
Formula Weight 297.91
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-2-iodoaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Bromo-2-iodoaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.