missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzoic acid, 97%

Product Code. 15559343 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15559343

Brand: Thermo Scientific Acros 443400250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68837-59-2 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68837-59-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
InChI Key RVCJOGNLYVNRDN-UHFFFAOYSA-N
Synonym 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5
PubChem CID 99570
IUPAC Name 4-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Br)C(=O)O

Specifications

Melting Point 182.0°C to 184.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzoic acid
compliance-icons
Product Identifier
  • 4-Bromo-2-methylbenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.