missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbiphenyl, 98%

Product Code. 11394805
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394805 5 g 5g
11384805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394805 Supplier Thermo Scientific Alfa Aesar Supplier No. L17875.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5002-26-6 | C13H11Br | 247.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5002-26-6
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD03093069
InChI Key ZBNARPVMXYNXQQ-UHFFFAOYSA-N
Synonym 4-bromo-2-methylbiphenyl, 4-bromo-2-methyl-1,1'-biphenyl, acmc-1asrj, 2-methyl-4-bromobiphenyl, 4-bromo-2-methyl-biphenyl, 3-methyl-4-phenylbromobenzene, 4-bromanyl-2-methyl-1-phenyl-benzene
PubChem CID 9964845
IUPAC Name 4-bromo-2-methyl-1-phenylbenzene
SMILES CC1=C(C=CC(=C1)Br)C2=CC=CC=C2

Specifications

Boiling Point 113°C to 114°C (0.7 mmHg)
Refractive Index 1.621
Quantity 5 g
Beilstein 1908937
Formula Weight 247.14
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.