missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrotoluene, 98%, Thermo Scientific™

Product Code. 10767393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10767393 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10767393 Supplier Thermo Scientific Acros Supplier No. 276210010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60956-26-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00041243
InChI Key KZNXALJXBRSMFL-UHFFFAOYSA-N
Synonym 4-bromo-2-nitrotoluene, 2-nitro-4-bromotoluene, 4-bromo-6-nitrotoluene, bromo 4--2-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, 4-bromo-1-methyl-2-nitro-benzene, 4-brom-2-nitrotoluol, pubchem10403, maybridge1_002072
PubChem CID 123546
IUPAC Name 4-bromo-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)Br)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43.0°C to 46.0°C
Color Orange to Yellow
Boiling Point 130.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 216.03
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromo-2-nitrotoluene
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-nitrotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.