missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10337454
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10337454 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10337454 Supplier Thermo Scientific Maybridge Supplier No. TL00627EA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 175278-09-8
CAS Min % 97.0
Density 1.6910g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5BrF3NO
MDL Number MFCD00179338
Quantity 10 g
Synonym 4-bromo-2-trifluoromethoxy aniline, 4-bromo-2-trifluoromethoxy-phenylamine, 2-amino-5-bromo trifluoromethoxy benzene, benzenamine, 4-bromo-2-trifluoromethoxy, 1-amino-4-bromo-2-trifluoromethoxy benzene, buttpark 50\01-74, 4-bromo-2-trifluoromethoxy phenylamine, pubchem2930, acmc-1c9j8, ksc184e5f
InChI Key QVILSWLYJYMGRN-UHFFFAOYSA-N
SMILES NC1=CC=C(Br)C=C1OC(F)(F)F
IUPAC Name 4-bromo-2-(trifluoromethoxy)aniline
Molecular Weight (g/mol) 256.02
PubChem CID 737382
Formula Weight 256.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.