missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)anisole, 98%

Product Code. 15495387
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495387 5 g 5g
15485387 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495387 Supplier Thermo Scientific Alfa Aesar Supplier No. H32645.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 853771-88-7 | C8H6BrF3O2 | 271.03 g/mol

4-Bromo-2-(trifluoromethoxy)anisole employed in biological evaluation of some analogs of the antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-(trifluoromethoxy)anisole employed in biological evaluation of some analogs of the antitumor agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 853771-88-7
Molecular Formula C8H6BrF3O2
Molecular Weight (g/mol) 271.03
MDL Number MFCD09907950
InChI Key QVGIROOBNUFIKC-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy anisole, 4-bromo-1-methoxy-2-trifluoromethoxy benzene, benzene,4-bromo-1-methoxy-2-trifluoromethoxy
PubChem CID 45790939
IUPAC Name 4-bromo-1-methoxy-2-(trifluoromethoxy)benzene
SMILES COC1=C(OC(F)(F)F)C=C(Br)C=C1

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 271.04
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)anisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.