missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)phenol, 98%, Thermo Scientific Chemicals

Product Code. 15484347
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484347 5 g 5g
15474347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484347 Supplier Thermo Scientific Alfa Aesar Supplier No. H31726.06
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

4-Bromo-2-(trifluoromethoxy)phenol are the basic building blocks numerous pharmaceutical drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-(trifluoromethoxy)phenol are the basic building blocks numerous pharmaceutical drugs.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 690264-39-2
Molecular Formula C7H4BrF3O2
Molecular Weight (g/mol) 257.01
MDL Number MFCD09037798
InChI Key GYXNTPVMXGBRQW-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy phenol, 4-bromo-2-trifluoromethoxy-phenol, 4-bromanyl-2-trifluoromethyloxy phenol, phenol, 4-bromo-2-trifluoromethoxy
PubChem CID 11780151
IUPAC Name 4-bromo-2-(trifluoromethoxy)phenol
SMILES OC1=C(OC(F)(F)F)C=C(Br)C=C1

Specifications

Melting Point 51°C to 55°C
Quantity 5 g
Formula Weight 257.01
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.