missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Product Code. 10341630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10341630 5 g Glass bottle 5g
10088089 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341630 Supplier Thermo Scientific Acros Supplier No. 438230050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50824-04-9
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
InChI Key PDPGERGWEOJVDC-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d
PubChem CID 142784
IUPAC Name 4-bromo-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 84°C to 88°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 241.01
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.