missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-difluorophenol, 99%, Thermo Scientific Chemicals

Product Code. 11386519
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11386519

Brand: Thermo Scientific Alfa Aesar H26153.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Bromo-3,5-difluorophenol is used as intermediates of Liquid Crystals. Reactant in the synthesis of difluoroethyleneoxy group liquid crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3,5-difluorophenol is used as intermediates of Liquid Crystals. Reactant in the synthesis of difluoroethyleneoxy group liquid crystal.

Solubility
Sparingly Soluble (8.2E-5 g/L) (25°C).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130191-91-2
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD08062385
InChI Key HHGOLZGZHXELSW-UHFFFAOYSA-N
PubChem CID 7172027
IUPAC Name 4-bromo-3,5-difluorophenol
SMILES C1=C(C=C(C(=C1F)Br)F)O

Specifications

Melting Point 72°C to 78°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym 2,6-difluoro-4-hydroxybromobenzene, phenol, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluoro-phenol, pubchem14520, intermediates-zcf02006, acmc-20a41x, 3,5-difluoro-4-bromophenol, phenol,4-bromo-3,5-difluoro
Solubility Information Sparingly Soluble (8.2E-5g/L) (25°C).
Formula Weight 208.99
Percent Purity 99%
Chemical Name or Material 4-Bromo-3,5-difluorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.