missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethoxybenzoic acid, 98%

Product Code. 11419447
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419447 5 g 5g
11429447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419447 Supplier Thermo Scientific Alfa Aesar Supplier No. B21449.06
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 56518-42-4 | C9H8BrO4 | 260.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56518-42-4
Molecular Formula C9H8BrO4
Molecular Weight (g/mol) 260.06
MDL Number MFCD01632140
InChI Key JNFZULSIYYVRJO-UHFFFAOYSA-M
Synonym benzoic acid, 4-bromo-3,5-dimethoxy, zlchem 1334, acmc-1az7r, jnfzulsiyyvrjo-uhfffaoysa, 3,5-dimethoxy-4-bromobenzoic acid, 4-bromo-3,5-dimethoxybenzoic acid
PubChem CID 2774026
IUPAC Name 4-bromo-3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1Br)C([O-])=O

Specifications

Melting Point 254°C to 258°C
Quantity 5 g
Formula Weight 261.07
Percent Purity 98%
Chemical Name or Material 4-Bromo-3,5-dimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.