missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylphenol, 99%

Product Code. 11934681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11934681 500 g 500g
10449780 10 g 10g
10459780 25 g 25g
10269513 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11934681 Supplier Thermo Scientific Acros Supplier No. 106875000
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7463-51-6 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7463-51-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00002315
InChI Key WMUWDPLTTLJNPE-UHFFFAOYSA-N
Synonym 4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
PubChem CID 81970
IUPAC Name 4-bromo-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Br)C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 113°C to 116°C
Color Colorless to Pink
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula BrC6H2(CH3)2OH
Quantity 500 g
Beilstein 06,I,244
Formula Weight 201.06
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material 4-Bromo-3, 5-dimethylphenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3,5-dimethylphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.