missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylphenol, 99%

Product Code. 11470267
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470267 5 g 5g
11480267 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470267 Supplier Thermo Scientific Alfa Aesar Supplier No. B21684.06
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 7463-51-6 | C8H9BrO | 201.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7463-51-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00002315
InChI Key WMUWDPLTTLJNPE-UHFFFAOYSA-N
Synonym 4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
PubChem CID 81970
IUPAC Name 4-bromo-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Br)C)O

Specifications

Melting Point 113°C to 117°C
Odor Phenol-like
Quantity 5 g
Beilstein 2207127
Formula Weight 201.07
Percent Purity 99%
Chemical Name or Material 4-Bromo-3,5-dimethylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.