missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzeneboronic acid, 95%

Product Code. 11365025
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365025 250 mg 250mg
11355025 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365025 Supplier Thermo Scientific Alfa Aesar Supplier No. L18514.MD
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 374790-97-3 | C6H5BBrFO2 | 218.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374790-97-3
Molecular Formula C6H5BBrFO2
Molecular Weight (g/mol) 218.82
MDL Number MFCD02093066
InChI Key ACLQPRPXJMWADE-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorobenzeneboronic acid, 4-bromo-3-fluorophenyl boronic acid, boronic acid, 4-bromo-3-fluorophenyl, boronic acid, b-4-bromo-3-fluorophenyl, pubchem7779, 4-bromo-3-fluoro-phenyl boronic acid, acmc-209it9, 4-bromo-3-fluorobenzeneboronicacid
PubChem CID 2773335
IUPAC Name (4-bromo-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Br)C(F)=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Formula Weight 218.82
Percent Purity 95%
Chemical Name or Material 4-Bromo-3-fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.