missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitroanisole, 96%

Product Code. 11317066
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317066 5 g 5g
11327066 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317066 Supplier Thermo Scientific Alfa Aesar Supplier No. L06944.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5344-78-5 | C7H6BrNO3 | 232.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5344-78-5
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD00051511
InChI Key KCOBIBRGPCFIGF-UHFFFAOYSA-N
Synonym 4-bromo-3-nitroanisole, 4-bromo-3-nitroanisol, benzene, 1-bromo-4-methoxy-2-nitro, 3-nitro-4-bromoanisole, 4-methoxy-2-nitrobromobenzene, 4-bromo-3-nitrothioanisole, 4-bromo-3-nitrophenyl methyl ether, 1-bromo-4-methoxy-2-nitro-benzene, timtec-bb sbb009974, pubchem2655
PubChem CID 79288
IUPAC Name 1-bromo-4-methoxy-2-nitrobenzene
SMILES COC1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 29°C to 32°C
Boiling Point 153°C to 154°C (13 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2446617
Formula Weight 232.03
Percent Purity 96%
Chemical Name or Material 4-Bromo-3-nitroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.