missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-iodobiphenyl, 98%

Product Code. p-7055707
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15461519

Brand: Thermo Scientific Alfa Aesar H64355.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105946-82-5 | C12H8BrI | 359.004 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105946-82-5
Molecular Formula C12H8BrI
Molecular Weight (g/mol) 359.004
MDL Number MFCD17676204
InChI Key GWOAJJWBCSUGHH-UHFFFAOYSA-N
Synonym 4-bromo-4'-iodobiphenyl, 4-bromo-4'-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-iodo, 1-bromo-4-4-iodophenyl benzene, 4'-bromo-4-iodobiphenyl, 4-iodo-4'-bromobiphenyl, 4-bromo-4'-iodo-biphenyl, 4-bromo-4/'-iodobiphenyl, ksc164g2f
PubChem CID 10904452
IUPAC Name 1-bromo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)Br

Specifications

Melting Point 175°C to 179°C (literature)
Quantity 25 g
UN Number UN3152
Beilstein 2518914
Sensitivity Light sensitive
Formula Weight 359
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-4'-iodobiphenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.