missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-methylbiphenyl, 97%, Thermo Scientific™

Product Code. 11490080
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490080 250 mg 250mg
11400090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490080 Supplier Thermo Scientific Alfa Aesar Supplier No. H50596.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromo-4′-methylbiphenyl is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50670-49-0
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.14
MDL Number MFCD00466236
InChI Key MYWLXURJCXISCT-UHFFFAOYSA-N
Synonym 4-bromo-4'-methylbiphenyl, 4-bromo-4'-methyl-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-methyl, biphenyl, 4-bromo-4'-methyl, 4-bromo-4'-methyl-biphenyl, 1-bromo-4-4-methylphenyl benzene, 1-4-bromophenyl-4-methylbenzene, 4-brromo-4-methylbiphenyl, 4-brom-4'-methyl-biphenyl, 4'-bromo-4-methyl-biphenyl
PubChem CID 521291
IUPAC Name 1-bromo-4-(4-methylphenyl)benzene
SMILES CC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 136°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 247.14
Percent Purity 97%
Chemical Name or Material 4-Bromo-4'-methylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.