missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-tert-butylbiphenyl, 98%

Product Code. 11436140
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436140 1 g 1g
11446140 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436140 Supplier Thermo Scientific Alfa Aesar Supplier No. H27076.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 162258-89-1 | C16H17Br | 289.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162258-89-1
Molecular Formula C16H17Br
Molecular Weight (g/mol) 289.22
MDL Number MFCD01321141
InChI Key QYNWFBYWVPMMRL-UHFFFAOYSA-N
Synonym 4-bromo-4'-tert-butylbiphenyl, 1,1'-biphenyl,4-bromo-4'-1,1-dimethylethyl, 4-bromo-4'-tert-butyl-1,1'-biphenyl, acmc-209do3, 4-bromo-4'-tertbutylbiphenyl, 4'-bromo-4-tert-butyl-biphenyl, 4-bromo-4/'-tert-butylbiphenyl, 4-bromo-4-tert-butylbiphenyl, 1-4-bromophenyl-4-tert-butylbenzene
PubChem CID 9971389
SMILES CC(C)(C)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 140°C to 143°C
Boiling Point 152°C (2 mmHg)
Quantity 1 g
IUPAC Name 4-bromo-4'-tert-butyl-1,1'-biphenyl
Formula Weight 289.21
Percent Purity 98%
Chemical Name or Material 4-Bromo-4'-tert-butylbiphenyl
compliance-icons
Product Identifier
  • 4-Bromo-4'-tert-butylbiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Bromo-4-tert-butylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.