missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulfonamide, 97%, Thermo Scientific™

Product Code. p-7037791
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462796 250 mg 250mg
15452796 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462796 Supplier Thermo Scientific Alfa Aesar Supplier No. H56605.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346721-67-3
Molecular Formula C14H13BrClNO4S
Molecular Weight (g/mol) 406.675
MDL Number MFCD00588020
InChI Key WBKSJEMNFRXWFA-UHFFFAOYSA-N
Synonym 4-bromo-n-4-chloro-2,5-dimethoxyphenyl benzenesulfonamide, 4-bromo-n-4-chloro-2,5-dimethoxyphenyl benzene-1-sulfonamide, cambridge id 5268579, 4-bromo-n-4-chloro-2,5-dimethoxy-phenyl-benzenesulfonamide, 4-bromo-n-4-chloro-2,5-dimethoxyphenyl-1-benzenesulfonamide, 4-bromo-n-4-chloro-2,5-bis methyloxy phenyl benzenesulfonamide
PubChem CID 1048928
IUPAC Name 4-bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulfonamide
SMILES COC1=CC(=C(C=C1NS(=O)(=O)C2=CC=C(C=C2)Br)OC)Cl

Specifications

Quantity 250 mg
Formula Weight 406.67
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.