missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-o-phenylenediamine, 97%

Product Code. 11451510
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451510 100 g 100g
11431510 5 g 5g
11441510 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451510 Supplier Thermo Scientific Alfa Aesar Supplier No. H26929.22
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1575-37-7 | C6H7BrN2 | 187.04 g/mol

4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1575-37-7
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD02660622
InChI Key WIHHVKUARKTSBU-UHFFFAOYSA-N
Synonym 4-bromo-1,2-benzenediamine, 4-bromo-1,2-diaminobenzene, 4-bromo-o-phenylenediamine, 1,2-diamino-4-bromobenzene, 2-amino-4-bromoaniline, 4-bromo-1,2-phenylenediamine, 4-bromo-2-aminoaniline, 1,2-benzenediamine, 4-bromo, 2-amino-4-bromophenylamine, 4-bromophenylene-1,2-diamine
PubChem CID 323593
IUPAC Name 4-bromobenzene-1,2-diamine
SMILES NC1=CC=C(Br)C=C1N

Specifications

Melting Point 64°C to 66°C
Quantity 100 g
UN Number UN2811
Solubility Information Soluble in chloroform.
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 4-Bromo-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.